siRNA Details
Gene Symbol: Gm20752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040750.1 | 5 | 2289 | |||
GenBank mRNA | AK158712.1 | - | 5 | 2291 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK143050.1 | - | 1068 | Chr. - - [-] on Build |
