siRNA Details
Gene Symbol: Gm34312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048295.1 | - | 2 | 279 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB648055.1 | - | 2 | 279 | Chr. 6 - 87597747 - 87598765 [+] on Build GRCm38 |
