siRNA Details
Gene Symbol: 4930404A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040495.1 | 2 | 213 | |||
GenBank mRNA | AK015072.1 | - | 2 | 214 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714600.1 | - | 214 | Chr. - - [-] on Build |
