siRNA Details
Gene Symbol: 4930404A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040495.1 | 1 | 63 | |||
GenBank mRNA | AK015072.1 | - | 1 | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714600.1 | - | 64 | Chr. - - [-] on Build |
