siRNA Details
Gene Symbol: 2700008L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012223.1 | - | 1 | 595 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB280877.1 | - | 1 | 168 | Chr. 11 - 70571146 - 70571340 [+] on Build GRCm38 |
