siRNA Details
Gene Symbol: Gm11418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132715.1 | - | 3 | 321 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614803.1 | - | 3 | 321 | Chr. 11 - 81587359 - 81593480 [+] on Build GRCm38 |
