siRNA Details
Gene Symbol: Pitpnm2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045369.1 | 4, 5 | 845 | |||
GenBank mRNA | AK016253.1 | - | 4, 5 | 844 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715864.1 | - | 4, 5 | 844 | Chr. 5 - 124229725 - 124236252 [+] on Build GRCm38 |
