siRNA Details
Gene Symbol: Grip1os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015613.1 | - | 3 | 540 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715164.1 | - | 3 | 540 | Chr. 10 - 119825478 - 119852390 [-] on Build GRCm38 |
