siRNA Details
Gene Symbol: Gm13599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029730.1 | - | 4 | 3878 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV266326.2 | - | 4 | 672 | Chr. 2 - 67309659 - 67396478 [-] on Build GRCm38 |
