siRNA Details
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032418.1 | - | 2680 | |||
AK085429.1 | - | 107 | ||||
AK162043.1 | - | 442 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032418.1 | - | 2680 | |||
AK085429.1 | - | 107 | ||||
AK162043.1 | - | 442 |