siRNA Details
Gene Symbol: Gm12603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144910.1 | - | 3 | 398 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734595.1 | - | 3 | 396 | Chr. 4 - 89050308 - 89084511 [+] on Build GRCm38 |
