siRNA Details
Gene Symbol: B230217C12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018341.1 | - | 2 | 1265 | ||
AK031146.1 | - | 1264 | ||||
AK035650.1 | - | 1247 | ||||
AK042827.1 | - | 1267 | ||||
AK080808.1 | - | 1264 | ||||
AK081899.1 | - | 1285 |
