siRNA Details
Gene Symbol: E130018N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040327.1 | 2 | 411 | |||
GenBank mRNA | AK018699.1 | - | 2 | 376 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E130018N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040327.1 | 2 | 411 | |||
GenBank mRNA | AK018699.1 | - | 2 | 376 |