siRNA Details
Gene Symbol: E130018N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040327.1 | 1 | 99 | |||
GenBank mRNA | AK018699.1 | - | 1 | 64 | ||
CJ177784.1 | - | 1 | 100 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E130018N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040327.1 | 1 | 99 | |||
GenBank mRNA | AK018699.1 | - | 1 | 64 | ||
CJ177784.1 | - | 1 | 100 |