siRNA Details
Gene Symbol: 2610027K06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077059.1 | 3 | 414 | |||
GenBank mRNA | AK085522.1 | - | 3 | 238 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610027K06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077059.1 | 3 | 414 | |||
GenBank mRNA | AK085522.1 | - | 3 | 238 |