siRNA Details
Gene Symbol: Gm19710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045749.1 | 2 | 200 | |||
GenBank mRNA | AK140585.1 | - | 2 | 202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB646753.1 | - | 2 | 202 | Chr. 3 - 89998758 - 90001275 [+] on Build GRCm38 |
