siRNA Details
Gene Symbol: 4930543N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016032.1 | - | 3 | 302 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715612.1 | - | 3 | 302 | Chr. 7 - 138554242 - 138566766 [+] on Build GRCm38 |
