siRNA Details
Gene Symbol: 4930543I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016030.1 | - | 1 | 853 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV269527.1 | - | 1 | 163 | Chr. 10 - 67786422 - 67786695 [+] on Build GRCm38 |
