siRNA Details
Gene Symbol: Gm15417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040403.1 | 2 | 903 | |||
NR_040404.1 | 2 | 364 | ||||
GenBank mRNA | AK136654.1 | - | 2 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721031.1 | - | 2 | 361 | Chr. 3 - 89394293 - 89398530 [+] on Build GRCm38 |
