siRNA Details
Gene Symbol: Msantd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013318.1 | - | 1 | 114 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK639586.1 | - | 1 | 220 | Chr. 9 - 37515446 - 37516142 [+] on Build GRCm38 |
