siRNA Details
Gene Symbol: 5430427M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045858.1 | 3 | 258 | |||
GenBank mRNA | AK017349.1 | - | 3 | 259 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717163.1 | - | 3 | 259 | Chr. 12 - 91045848 - 91056369 [+] on Build GRCm38 |
