siRNA Details
Gene Symbol: 2610028D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011582.1 | - | 1 | 1202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY754712.1 | - | 1 | 171 | Chr. 9 - 45797352 - 45798029 [-] on Build GRCm38 |
