siRNA Details
Gene Symbol: 9430087D07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020510.1 | - | 1 | 224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721899.1 | - | 1 | 224 | Chr. 1 - 15570583 - 15571221 [+] on Build GRCm38 |
