siRNA Details
Gene Symbol: 9430087N24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020513.1 | - | 1 | 236 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721905.1 | - | 1 | 236 | Chr. 15 - 43734088 - 43734736 [-] on Build GRCm38 |
