siRNA Details
Gene Symbol: 1700024N05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006308.1 | - | 1 | 111 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706384.1 | - | 1 | 109 | Chr. 6 - 43162454 - 43172316 [-] on Build GRCm38 |
