siRNA Details
Gene Symbol: 1700024I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006304.1 | - | 5 | 616 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706381.1 | - | 5 | 613 | Chr. 13 - 63675768 - 63734298 [-] on Build GRCm38 |
