siRNA Details
Gene Symbol: 1700024K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006306.1 | - | 1 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN839877.1 | - | 1 | 329 | Chr. 9 - 99872261 - 99872674 [-] on Build GRCm38 |
