siRNA Details
Gene Symbol: Gm26679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083202.1 | - | 3 | 518 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB653326.1 | - | 3 | 518 | Chr. 14 - 120384859 - 120388985 [-] on Build GRCm38 |
