siRNA Details
Gene Symbol: 1700020K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006169.1 | - | 1 | 429 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706270.1 | - | 1 | 426 | Chr. 10 - 63800871 - 63801353 [+] on Build GRCm38 |
