siRNA Details
Gene Symbol: 1700020G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006160.1 | - | 1 | 48 | ||
AK015597.1 | - | 1 | 34 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU939460.1 | - | 1 | 35 | Chr. 10 - 110812804 - 110896535 [-] on Build GRCm38 |
