siRNA Details
Gene Symbol: 1700020G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006160.1 | - | 3 | 358 | ||
AK015597.1 | - | 3 | 335 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700020G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006160.1 | - | 3 | 358 | ||
AK015597.1 | - | 3 | 335 |