siRNA Details
Gene Symbol: 1700020G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045979.1 | 1 | 26 | |||
GenBank mRNA | AK006160.1 | - | 1 | 70 | ||
AK076938.1 | - | 1 | 27 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700020G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045979.1 | 1 | 26 | |||
GenBank mRNA | AK006160.1 | - | 1 | 70 | ||
AK076938.1 | - | 1 | 27 |