siRNA Details
Gene Symbol: Gm26664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037841.1 | - | 2 | 630 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB160530.2 | - | 2 | 571 | Chr. 8 - 84665185 - 84669406 [+] on Build GRCm38 |
