siRNA Details
Gene Symbol: 2900055J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045177.1 | 1 | 297 | |||
GenBank mRNA | AK013700.1 | - | 1 | 298 | ||
BM932606.1 | - | 1 | 219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF535901.1 | - | 1 | 274 | Chr. 18 - 40256985 - 40257647 [+] on Build GRCm38 |
