siRNA Details
Gene Symbol: 2900055J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045177.1 | 1 | 347 | |||
GenBank mRNA | AK013700.1 | - | 1 | 348 | ||
BM932606.1 | - | 1 | 269 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2900055J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045177.1 | 1 | 347 | |||
GenBank mRNA | AK013700.1 | - | 1 | 348 | ||
BM932606.1 | - | 1 | 269 |