siRNA Details
Gene Symbol: Gm12610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166205.1 | - | 1167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB821837.1 | - | 1 | 114 | Chr. 4 - 89515259 - 89515685 [+] on Build GRCm38 |
