siRNA Details
Gene Symbol: Saxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076898.1 | - | 3 | 606 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV269471.2 | - | 3 | 627 | Chr. 4 - 86481549 - 86558328 [-] on Build GRCm38 |
