siRNA Details
Gene Symbol: Wdfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158235.1 | - | 390 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY753019.1 | - | 2 | 387 | Chr. 14 - 62837677 - 62838205 [-] on Build GRCm38 |
