siRNA Details
Gene Symbol: Gm26684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142516.1 | - | 2 | 591 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660796.1 | - | 2 | 586 | Chr. 15 - 54952952 - 54954113 [+] on Build GRCm38 |
