siRNA Details
Gene Symbol: Gm30025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147035.1 | - | 1 | 488 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY760904.1 | - | 1 | 433 | Chr. 12 - 80435262 - 80435931 [+] on Build GRCm38 |
