siRNA Details
Gene Symbol: D730044K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029389.1 | - | 1 | 1636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV250913.2 | - | 1 | 458 | Chr. 15 - 34444158 - 34444812 [+] on Build GRCm38 |
