siRNA Details
Gene Symbol: A830019L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040551.1 | 4 | 917 | |||
GenBank mRNA | AK043679.1 | - | 4 | 918 | ||
AK086499.1 | - | 552 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB663142.1 | - | 4 | 552 | Chr. 3 - 143242775 - 143257313 [+] on Build GRCm38 |
