siRNA Details
Gene Symbol: A830019L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040551.1 | 4 | 1242 | |||
GenBank mRNA | AK043679.1 | - | 4 | 1243 | ||
AK086499.1 | - | 877 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A830019L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040551.1 | 4 | 1242 | |||
GenBank mRNA | AK043679.1 | - | 4 | 1243 | ||
AK086499.1 | - | 877 |