siRNA Details
Gene Symbol: Gm16833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045754.1 | 1, 2 | 173 | |||
GenBank mRNA | AK143934.1 | - | 1, 2 | 173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB846505.1 | - | 1, 2 | 173 | Chr. 9 - 9236288 - 9260620 [+] on Build GRCm38 |
