siRNA Details
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U15218.1 | - | 1 | 148 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX568898.1 | - | 1 | 101 | Chr. X - 83855407 - 83887137 [+] on Build GRCm38 |
