siRNA Details
Gene Symbol: 4930571O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110476.1 | 1 | 57 | |||
GenBank mRNA | AK019805.1 | - | 1 | 58 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715894.1 | - | 1 | 58 | Chr. 8 - 113517282 - 113550810 [+] on Build GRCm38 |
