siRNA Details
Gene Symbol: Gm31938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077964.1 | - | 3 | 554 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB621333.1 | - | 3 | 550 | Chr. 12 - 4524115 - 4527964 [+] on Build GRCm38 |
