siRNA Details
Gene Symbol: 4933409D19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016750.1 | - | 2 | 481 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716430.1 | - | 2 | 481 | Chr. 1 - 136603965 - 136605468 [-] on Build GRCm38 |
