siRNA Details
Gene Symbol: 2310034P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009626.1 | - | 1 | 200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709290.1 | - | 1 | 196 | Chr. 7 - 83884915 - 83885193 [+] on Build GRCm38 |
