siRNA Details
Gene Symbol: 1700112D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007174.1 | - | 245 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707263.1 | - | 2 | 245 | Chr. 3 - 28738780 - 28780700 [-] on Build GRCm38 |
